missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CKAP4/p63 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11600-100
This item is not returnable.
View return policy
Description
CKAP4/p63 Monoclonal antibody specifically detects CKAP4/p63 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry,Immunocytochemistry
Specifications
| CKAP4/p63 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 63 kDa membrane protein, CLIMP-63, cytoskeleton-associated protein 4, ERGIC-63, MGC99554, P63, transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediatecompartment, type-II transmembrane protein p63 | |
| Mouse myeloma cell line, NS0-derived human CKAP-4, His128-Val602, Accession # Q07065 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry | |
| 1086526 | |
| Western Blot 1 μg/mL, Simple Western 4 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Q07065 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10970 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur