missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Claudin-17 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4619-SP
This item is not returnable.
View return policy
Description
Claudin-17 Monoclonal specifically detects Claudin-17 in Human samples. It is validated for Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Claudin-17 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P56750 | |
| CLDN17 | |
| NS0 mouse myeloma cell line transfected with human Claudin-17 Met1-Val224 Accession # P56750 | |
| 25 μg | |
| Primary | |
| Detects human Claudin-17. Stains human Claudin-17 transfectants but not irrelevant transfectants in flow cytometry. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 410509 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| claudin 17, Claudin17, CLDN17, human CLDN17 gene for claudin-17, 10claudin-17, MGC126552, MGC126554 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26285 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction