missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Claudin-4 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4219N-100UG
This item is not returnable.
View return policy
Description
Claudin-4 Monoclonal specifically detects Claudin-4 in Human samples. It is validated for Flow Cytometry.
Specifications
| Claudin-4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_001296 | |
| CLDN4 | |
| NS0 mouse myeloma cell line transfected with human Claudin-4 Met1-Val209 Accession # NP_001296 | |
| RUO | |
| 1364 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 382321 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| claudin 4, Claudin4, CLDN4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-R, CPE-RCPETR, CPE-receptor, CPETR1, hCPE-R, WBSCR8, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Claudin-4. Stains human Claudin-4 transfectants but not irrelevant transfectants. Recognizes an epitope in the first extracellular loop (aa Met29-Arg81). | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction