missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Claudin-8 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC5275V-100UG
This item is not returnable.
View return policy
Description
Claudin-8 Monoclonal specifically detects Claudin-8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Claudin-8 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P56748 | |
| CLDN8 | |
| HEK293 human embryonic kidney cell line transfected with human Claudin-8 Met1-Val225 (Ser151Pro) Accession # P56748 | |
| RUO | |
| 9073 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 474210 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| claudin 8, Claudin8, CLDN8, human CLDN8 gene for claudin-810claudin-8 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Claudin-8. Stains human Claudin-8 transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction