missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC2L Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems FAB10204S-100UG
This item is not returnable.
View return policy
Description
CLEC2L Monoclonal antibody specifically detects CLEC2L in Human samples. It is validated for Flow Cytometry
Specifications
| CLEC2L | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide | |
| C-type lectin domain family 2 member L, C-type lectin domain family 2, member L | |
| Human embryonic kidney cell, HEK293-derived human CLEC2L protein, Ser90-Thr214, Accession # P0C7M8-1 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2799A | |
| Flow Cytometry 0.5 ug/10^6 cells | |
| P0C7M8-1 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 154790 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction