missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC4D/CLECSF8 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2806
This item is not returnable.
View return policy
Description
CLEC4D/CLECSF8 Monoclonal specifically detects CLEC4D/CLECSF8 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| CLEC4D/CLECSF8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Q8WXI8 | |
| CLEC4D | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8 Gly52-Asn215 Accession # Q8WXI8 | |
| 100 μg | |
| Primary | |
| Detects human CLEC4D/CLECSF8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CLECSF9, rhCLECSF13, rhOCIL, or rhOCILrp2 is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 413512 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD368, CLEC6, CLEC-6, CLECSF8, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 338339 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction