missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CLEC9a Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6049
This item is not returnable.
View return policy
Description
CLEC9a Polyclonal specifically detects CLEC9a in Human samples. It is validated for Western Blot, Immunohistochemistry, Binding Inhibition.
Specifications
| CLEC9a | |
| Polyclonal | |
| Unconjugated | |
| Q6UXN8 | |
| CLEC9A | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC9a Lys57-Val241 Accession # Q6UXN8 | |
| 100 μg | |
| Primary | |
| Detects human CLEC9a in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse CLEC9a is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Binding Inhibition 10 ug/mL | |
| CD370, C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 283420 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction