missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Coilin (aa 428-576) Control Fragment Recombinant Protein

Product Code. 30210091
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30210091

Brand: Invitrogen™ RP105782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110950 (PA5-110950. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is an integral component of Cajal bodies (also called coiled bodies). Cajal bodies are nuclear suborganelles of varying number and composition that are involved in the post-transcriptional modification of small nuclear and small nucleolar RNAs. The N-terminus of the coilin protein directs its self-oligomerization while the C-terminus influences the number of nuclear bodies assembled per cell. Differential methylation and phosphorylation of coilin likely influences its localization among nuclear bodies and the composition and assembly of Cajal bodies. This gene has pseudogenes on chromosome 4 and chromosome 14.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P38432
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8161
Name Human Coilin (aa 428-576) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C79982; CLN80; COIL; Coilin; coilin p80; hypothetical protein LOC507580; LOW QUALITY PROTEIN: coilin; nuclear phosphoprotein of Cajal body; p80; p80-coilin; wu:fb08h11
Common Name Coilin
Gene Symbol COIL
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NRSTDNQRQQQLNDVVKNSSTIIQNPVETPKKDYSLLPLLAAAPQVGEKIAFKLLELTSSYSPDVSDYKEGRILSHNPETQQVDIEILSSLPALREPGKFDLVYHNENGAEVVEYAVTQESKITVFWKELIDPRLIIESPSNTSSTEPA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.