missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Collagen XXV alpha 1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF4317
This item is not returnable.
View return policy
Description
Collagen XXV alpha 1 Polyclonal specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Western Blot.
Specifications
| Collagen XXV alpha 1 | |
| Polyclonal | |
| Unconjugated | |
| NP_942014 | |
| COL25A1 | |
| Mouse myeloma cell line NS0-derived recombinant human Collagen XXV alpha 1 Gly189-Lys654 Accession # NP_942014 | |
| 100 μg | |
| Primary | |
| Detects human Collagen XXV alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human COL23A1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, COL25A1, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 84570 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction