Learn More
Human Complement Component C1s (Research Grade Sutimlimab Biosimilar) Antibody, Novus Biologicals™
Human Monoclonal Antibody
Brand:  R&D Systems MAB11489-100
 View more versions of this product
Product Code. 30130443
  • € 521.00 / 100µg
Description
Complement Component C1s Monoclonal antibody specifically detects Complement Component C1s in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complement Component C1s
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
basic proline-rich peptide IB-1, C1 esterase, C1S, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757
NS0 mouse myeloma cell line transfected with human CD38, Met1-Ile300, Accession # P28907
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG1
ELISA
Hu222
Flow Cytometry 0.25 μg/10^6 cells, ELISA
P28907
Human
Protein A or G purified from hybridoma culture supernatant
RUO
716