missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human COMT Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7386
This item is not returnable.
View return policy
Description
COMT Polyclonal specifically detects COMT in Human samples. It is validated for Western Blot.
Specifications
| COMT | |
| Polyclonal | |
| Unconjugated | |
| P21964 | |
| COMT | |
| E. coli-derived recombinant human COMT Gly52-Pro271 (Val158Met) Accession # P21964 | |
| 100 μg | |
| Primary | |
| Detects human COMT in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| catechol O-methyltransferase, catechol-O-methyltransferase, EC 2.1.1.6 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1312 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto