missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Connexin 43/GJA1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB7737G-100UG
This item is not returnable.
View return policy
Description
Connexin 43/GJA1 Monoclonal specifically detects Connexin 43/GJA1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Connexin 43/GJA1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P17302 | |
| GJA1 | |
| Mouse myeloma cell line NS0 transfected with human Connexin 43/GJA1 Accession # P17302 | |
| RUO | |
| 2697 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 578618 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| connexin 43, connexin-43, Cx43, CX43gap junction protein, alpha-like, DFNB38, Gap junction 43 kDa heart protein, gap junction alpha-1 protein, gap junction protein, alpha 1, 43kDa, gap junction protein, alpha 1, 43kDa (connexin 43), GJA1, GJAL, HSS, ODD, ODDD, ODOD, SDTY3 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Connexin 43/GJA1. Stains human Connexin 43/GJA1 transfectants but not irrelevant transfectants in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction