missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Contactin-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF904
This item is not returnable.
View return policy
Description
Contactin-1 Polyclonal specifically detects Contactin-1 in Human samples. It is validated for Western Blot.
Specifications
| Contactin-1 | |
| Polyclonal | |
| Biotin | |
| CAA79696 | |
| CNTN1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Contactin-1 Glu21-Ser993 Accession # CAA79696 | |
| 50 μg | |
| Primary | |
| Detects humanáContactin-1 in Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CNTN1, contactin 1, contactin-1, Contactin1, F11, F3, F3cam, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1272 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction