missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (23), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14657MFV610
This item is not returnable.
View return policy
Description
Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-229E, HCoV-NL63 samples. It is validated for Western Blot, ELISA
Specifications
| Human Coronavirus Spike Protein | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 3200426 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 23 | |
| Western Blot, ELISA | |
| HKU1, Spike glycoprotein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Coronavirus Spike S1+S2 ECD protein. | |
| 0.1 mL | |
| Primary | |
| HCoV-229E, HCoV-NL63 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction