missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CPNE1 (aa 222-251) Control Fragment Recombinant Protein

Product Code. 30205230
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205230

Brand: Invitrogen™ RP105570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84904 (PA5-84904. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Calcium-dependent membrane-binding proteins may regulate molecular events at the interface of the cell membrane and cytoplasm. This gene encodes a calcium-dependent protein that also contains two N-terminal type II C2 domains and an integrin A domain-like sequence in the C-terminus. However, the encoded protein does not contain a predicted signal sequence or transmembrane domains. This protein has a broad tissue distribution and it may function in membrane trafficking. This gene and the gene for RNA binding motif protein 12 overlap at map location 20q11.21. Alternate splicing results in multiple transcript variants encoding different proteins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99829
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8904
Name Human CPNE1 (aa 222-251) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810028N16Rik; Chromobindin 17; copine 1; copine I; copine III; copine III, like; copine-1; COPN1; CPN1; Cpne1; cpne3; cpne3l; MGC1142; mKIAA4108; OTTHUMP00000030775; OTTHUMP00000030776; Unknown (protein for IMAGE:8126132); wu:fb69d07; wu:fc45a04; zgc:55873
Common Name CPNE1
Gene Symbol CPNE1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DLIGTFHTSLAQLQAVPAEFECIHPEKQQK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.