missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRACC/SLAMF7 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1906V-100UG
This item is not returnable.
View return policy
Description
CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Human samples. It is validated for Flow Cytometry.
Specifications
| CRACC/SLAMF7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9NQ25 | |
| SLAMF7 | |
| Mouse myeloma cell line NS0-derived recombinant human CRACC/SLAMF7 Lys27-Ser225 Accession # Q9NQ25 | |
| RUO | |
| 57823 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 235614 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7, SLAMF7 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CRACC/SLAMF7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human NTB-AR is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction