Learn More
R&D Systems™ Human CRBN (NP_057386.2) VersaClone cDNA
Specifications
Specifications
| Gene | CRBN |
| Accession Number | NP_057386.2 |
| Gene ID (Entrez) | 51185 |
| Gene Alias | cereblon, DKFZp781K0715, MGC27358, MRT2A, non-syndromic, autosomal recessive, 2 A, protein cereblon, protein x 0001 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 10 μg |