missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRELD1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3128-SP
This item is not returnable.
View return policy
Description
CRELD1 Monoclonal specifically detects CRELD1 in Human samples. It is validated for Neutralization.
Specifications
| CRELD1 | |
| Monoclonal | |
| LYOPH | |
| Neutralization 0.15-0.75 ug/mL | |
| Q96HD1 | |
| CRELD1 | |
| Mouse myeloma cell line NS0-derived recombinant human CRELD-1 Gln30-Glu362 Accession # Q96HD1 | |
| 25 μg | |
| Primary | |
| Detects human CRELD-1 in ELISA. | |
| Human | |
| Purified |
| Neutralization | |
| 842314 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| atrioventricular septal defect 2, AVSD2, CIRRIN, cysteine-rich with EGF-like domain protein 1, cysteine-rich with EGF-like domains 1, DKFZp566D213 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 78987 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction