missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APP Antibody (OTI4C9), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70197AF488
This item is not returnable.
View return policy
Description
APP Monoclonal specifically detects APP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| APP | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| APP | |
| Full length recombinant protein of human APP (NP_000475) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI4C9 | |
| Western Blot, Immunohistochemistry | |
| amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP | |
| Mouse | |
| Affinity Purified | |
| Alzheimers Research, Apoptosis, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 351.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction