missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRHR1 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3930N-100UG
This item is not returnable.
View return policy
Description
CRHR1/CRF1 Monoclonal specifically detects CRHR1/CRF1 in Human samples. It is validated for Flow Cytometry.
Specifications
| CRHR1/CRF1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_004373 | |
| CRHR1 | |
| NS0 mouse myeloma cell line transfected with human CRHR-1 Ser24-Val415 Accession # NP_004373 | |
| RUO | |
| 1394 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 343919 | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CHR-R1, corticotropin releasing hormone receptor 1, corticotropin-releasing factor receptor 1, corticotropin-releasing factor type 1 receptor, Corticotropin-releasing hormone receptor 1, CRF1, CRFR, CRF-R, CRFR1, CRF-R1, CRF-R-1, CRFR-1, CRH-R-1, CRHR1f, CRH-R1h, CRHR1L, CRHRCRH-R1, seven transmembrane helix receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CRHR-1 transfectants but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction