missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cripto-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF145
This item is not returnable.
View return policy
Description
CRIPTO Polyclonal specifically detects CRIPTO in Human samples. It is validated for Western Blot.
Specifications
| CRIPTO | |
| Polyclonal | |
| Unconjugated | |
| P13385 | |
| TDGF1 | |
| E. coli-derived recombinant human Cripto-1 Arg38-Tyr188 Accession # P13385 | |
| 100 μg | |
| Primary | |
| Detects human Cripto-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse Cripto is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6997 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction