missing translation for 'onlineSavingsMsg'
Learn More

Human Cripto-1 APC-conjugated Antibody, R&D Systems™

Product Code. 18115331
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18115331 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18115331 Supplier R&D Systems Supplier No. FAB2772A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRIPTO Monoclonal specifically detects CRIPTO in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRIPTO
Applications Flow Cytometry
Classification Monoclonal
Clone 89633
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P13385
Gene Alias CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1
Gene Symbols TDGF1
Host Species Mouse
Immunogen E. coli-derived recombinant human Cripto-1 Arg38-Tyr188 Accession # P13385
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6997
Test Specificity Detects human Cripto-1 in ELISAs. In sandwich immunoassays, no cross-reactivity with recombinant mouse Cripto-1, recombinant human (rh) EGF, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 2, rhTGF-beta 3, or rhCryptic is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.