missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cripto Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2772S-100UG
This item is not returnable.
View return policy
Description
CRIPTO Monoclonal specifically detects CRIPTO in Human samples. It is validated for Flow Cytometry.
Specifications
| CRIPTO | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1 | |
| E. coli-derived recombinant human Cripto-1, Arg38-Tyr188, Accession # P13385 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 89633 | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| P13385 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6997 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction