missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRTAM Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF1695
This item is not returnable.
View return policy
Description
CRTAM Polyclonal specifically detects CRTAM in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| CRTAM | |
| Polyclonal | |
| Biotin | |
| O95727 | |
| CRTAM | |
| Mouse myeloma cell line NS0-derived recombinant human CRTAM Ser18-Ser286 Accession # O95727 | |
| 50 μg | |
| Primary | |
| Detects human CRTAM in ELISAs and Western blots. In sandwich immunoassays, less than 2% cross-reactivity with recombinant mouse CRTAM is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CD355, CD355 antigen, class I MHC restricted T cell associated molecule, class-I MHC-restricted T cell associated molecule, Class-I MHC-restricted T-cell-associated molecule, cytotoxic and regulatory T cell molecule, cytotoxic and regulatory T-cell molecule | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 56253 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction