missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CRX Antibody, R&D Systems™
Sheep Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF7085
This item is not returnable.
View return policy
Description
CRX/CORD2 Polyclonal specifically detects CRX/CORD2 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| CRX/CORD2 | |
| Polyclonal | |
| Unconjugated | |
| O43186 | |
| CRX | |
| E. coli-derived recombinant human CRX Ala166-Leu299 Accession # O43186 | |
| 100 μg | |
| Primary | |
| Detects human CRX in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| cone-rod homeobox, CORD2, CRDcone-rod homeobox protein, LCA7, LCA7orthodenticle homeobox 3, OTX3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1406 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction