missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CSRP3 (aa 76-129) Control Fragment Recombinant Protein

Product Code. 30181924
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181924

Brand: Invitrogen™ RP98423

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83620 (PA5-83620. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the CSRP family of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. The LIM/double zinc-finger motif found in this protein is found in a group of proteins with critical functions in gene regulation, cell growth, and somatic differentiation. Mutations in this gene are thought to cause heritable forms of hypertrophic cardiomyopathy (HCM) and dilated cardiomyopathy (DCM) in humans. Alternatively spliced transcript variants with different 5' UTR, but encoding the same protein, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P50461
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8048
Name Human CSRP3 (aa 76-129) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias cardiac LIM domain protein; cardiac LIM protein; Clp; CMD1M; CMH12; CRP3; Csrp3; cysteine and glycine rich protein 3; cysteine and glycine-rich protein 3; cysteine and glycine-rich protein 3 (cardiac LIM protein); cysteine-rich protein 3; LIM domain only 4; LIM domain protein, cardiac; LMO4; MGC14488; MGC61993; MLP; MMLP; Muscle LIM protein; muscle lim protein isoform
Common Name CSRP3
Gene Symbol Csrp3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GAGCLSTDTGEHLGLQFQQSPKPARSVTTSNPSKFTAKFGESEKCPRCGKSVYA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.