missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTLA-4 Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC3251V-100UG
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CTLA-4 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CTLA-4 Ala37-Phe162 Accession # P16410 | |
| 100 μg | |
| 1493 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 922101 | |
| Intracellular Staining by Flow Cytometry 0.25-1μg/106 cells | |
| P16410 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CTLA-4 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction