missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTLA-4 PE-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB386P
This item is not returnable.
View return policy
Description
CTLA-4 Polyclonal specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry.
Specifications
| CTLA-4 | |
| Polyclonal | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1493 | |
| Human | |
| IgG |
| Flow Cytometry | |
| R-PE | |
| Q6GR94 | |
| CTLA4 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human CTLA-4 Ala37-Phe162 Accession # Q6GR94 | |
| 100 Tests | |
| Primary | |
| Detects human CTLA-4 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse CTLA-4 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction