missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTRP4/C1qTNF4 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF3456
This item is not returnable.
View return policy
Description
C1qTNF4/CTRP4 Polyclonal specifically detects C1qTNF4/CTRP4 in Human samples. It is validated for Western Blot.
Specifications
| C1qTNF4/CTRP4 | |
| Polyclonal | |
| Biotin | |
| Q9BXJ3 | |
| C1QTNF4 | |
| Mouse myeloma cell line NS0-derived recombinant human CTRP4/C1qTNF4 Leu17-Leu329 Accession # Q9BXJ3 | |
| 50 μg | |
| Primary | |
| Detects human CTRP4/C1qTNF4 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant human (rh) CTRP5 is observed and less than 1% cross-reactivity with rhCTRP1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| C1q and tumor necrosis factor related protein 4, C1qTNF4, complement C1q tumor necrosis factor-related protein 4, complement-c1q tumor necrosis factor-related protein 4, CTRP4ZACRP4, ZACRP4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 114900 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction