Learn More
Human CX3CL1/Fractalkine Chemokine Domain Biotinylated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems BAM365
Product Code. 18160891
  • € 560.00 / 100µg
Description
CX3CL1/Fractalkine Monoclonal specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CX3CL1/Fractalkine
Monoclonal
LYOPH
Western Blot 1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
P78423
CX3CL1
NS0-derived and E.coli-derived Recombinant Human Fractalkine Gln25-Arg339 Accession # P78423
100 μg
Primary
Detects human CX3CL1 in ELISAs and Western blots. In ELISAs, this antibody shows approximately 8% cross-reactivity with recombinant mouse (rm) CXCL6 and less than 0.2% cross-reactivity with rhCXCL6, rhCXCL9, and rrCX3CL1.
Human
Purified
Western Blot, ELISA
51637
Biotin
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1
Mouse
Protein A or G purified from ascites
RUO
6376
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.