missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL1/GRO alpha/KC/CINC-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AB-275-PB
This item is not returnable.
View return policy
Description
CXCL1/GRO alpha/KC/CINC-1 Polyclonal specifically detects CXCL1/GRO alpha/KC/CINC-1 in Human samples. It is validated for Western Blot.
Specifications
| CXCL1/GRO alpha/KC/CINC-1 | |
| Polyclonal | |
| Unconjugated | |
| P09341 | |
| CXCL1 | |
| E. coli-derived recombinant human CXCL1/GRO alpha Ala35-Asn107 Accession # P09341 | |
| 1 mg | |
| Primary | |
| Detects human GRO alpha in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 30% cross-reactivity with recombinant human (rh) GRO beta , rhGRO gamma , and rmMIP-2, 20% cross-reactivity with rrCINC-1, and 10% cross-reactivity with rrCINC-2 alpha , rrCINC-2 beta , rmKC, rhIL-8, and rhNAP-2. | |
| Human | |
| Purified |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1 | |
| Goat | |
| Protein A or G purified | |
| RUO | |
| 2919 | |
| Reconstitute at 1 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction