missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL10/IP-10/CRG-2 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 15 publications
Brand: R&D Systems BAF266
This item is not returnable.
View return policy
Description
CXCL10/IP-10/CRG-2 Polyclonal specifically detects CXCL10/IP-10/CRG-2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, ELISA Detection (Matched Antibody Pair).
Specifications
| CXCL10/IP-10/CRG-2 | |
| Polyclonal | |
| Biotin | |
| C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3627 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Flow Cytometry, ELISA | |
| LYOPH | |
| Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CXCL10 | |
| E. coli-derived recombinant human CXCL10/IP-10 | |
| 50 μg | |
| Primary | |
| Detects human CXCL10/IP-10/CRG-2 in ELISAs and Western blots. In sandwich ELISAs, less than 0.1% cross-reactivity with recombinant mouse CRG-2, recombinant human (rh) Lymphotactin, rhGRO alpha , and rhMIG is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction