missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CXCL12 (SDF-1a), Animal-Free Recombinant Protein, PeproTech®

Product Code. 17806458
Click to view available options
Quantity:
2 μg
250 μg
50 μg
Unit Size:
250µg
2µg
50µg
This item is not returnable. View return policy

Product Code. 17806458

Brand: Gibco™ AF30028A2UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human SDF-1alpha is an 8.0 kDa protein containing 68 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48061
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 6387
Molecular Weight (g/mol) 8 kDa
Name Human CXCL12 (SDF-1 A), Animal-Free
Purification Method HPLC, SDS-PAGE
Quantity 2 μg
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; Cxcl12; H-SDF-1-alpha; Intercrine reduced in hepatomas; IRH; Pbsf; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; SDF 1 A; SDF 1 alpha; SDF 1 A; SDF1; SDF-1; SDF1 alpha; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); SDF1a; Stromal cell-derived factor 1; stromal cell-derived factor-1 alpha; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tpar1
Common Name CXCL12 alpha
Gene Symbol CXCL12
Biological Activity Determined by its ability to chemoattract IL-2-activated human T cells using a concentration range of 20.0-80.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNK
Content And Storage -20°C
Activity Determined by its ability to chemoattract IL-2-activated human T cells using a concentration range of 20.0-80.0 ng/ml
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.