missing translation for 'onlineSavingsMsg'
Learn More

Human CXCL13/BLC/BCA-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18760147
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18760147 25 μg 25µg
18753697 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18760147 Supplier R&D Systems Supplier No. MAB8012SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

CXCL13/BLC/BCA-1 Monoclonal specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Immunohistochemistry, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CXCL13/BLC/BCA-1
Applications Flow Cytometry, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 53602
Conjugate Unconjugated
Dilution Flow Cytometry 10 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL, Neutralization 0.5-2.5 ug/mL
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. O43927
Gene Alias ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL13/BLC/BCA-1, Val23-Arg94, Accession # O43927
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10563
Test Specificity Detects human CXCL13/BLC/BCA-1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.