missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human CXCL13/BLC/BCA-1 APC-conjugated Antibody, Novus Biologicals™

Mouse Monoclonal Antibody
Brand: Novus Biologicals™ IC8012A-100
This item is not returnable.
View return policy
Description
CXCL13/BLC/BCA-1 Monoclonal antibody specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
| CXCL13/BLC/BCA-1 | |
| Monoclonal | |
| APC | |
| Supplied as a 0.2 μm filtered solution in Tris, NaCl and Sodium Azide | |
| ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
| E. coli -derived recombinant human CXCL13/BLC/BCA-1, Val23-Arg94, Accession # O43927 | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining) | |
| 53602 | |
| Intracellular Staining by Flow Cytometry 10 μL/106 cells | |
| O43927 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10563 | |
| Protect from light.Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction