missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CXCL13/BLC/BCA-1 Quantikine ELISA Kit

Product Code. 18721964
missing translation for 'orderingAttributeHoverText'
Sufficient For:
1 x 96-well Plate
missing translation for 'unitSize'
1 set
This item is not returnable. View return policy

Product Code. 18721964

missing translation for 'mfr': R&D Systems™ DCX130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CXCL13/BLC/BCA-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CXCL13/BLC/BCA-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CXCL13/BLC/BCA-1 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), saliva (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 7.80 - 500pg/mL
Assay Sensitivity 3.97pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Saliva, Serum
Specificity Natural and Recombinant Human BLC/BCA-1
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 10563
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),EDTA Plasma (50uL),Heparin Plasma (50uL),Saliva (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1, BCA-1, BCA1B-cell chemoattractant, BCA-1 CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLC, BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.