missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL17/VCC-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4207U-100UG
This item is not returnable.
View return policy
Description
CXCL17/VCC-1 Monoclonal specifically detects CXCL17/VCC-1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CXCL17/VCC-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q6UXB2 | |
| CXCL17 | |
| E. coli-derived recombinant human CXCL17/VCC-1 Leu24-Leu119 Accession # Q6UXB2 | |
| RUO | |
| 284340 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 422208 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Chemokine (C-X-C Motif) Ligand 17, C-X-C Motif Chemokine 17, CXCL17, Dcip1, Dendritic Cell And Monocyte Chemokine-Like Protein, DMC, UNQ473, VCC1, VCC-1, VEGF Coregulated Chemokine 1, VEGF Co-Regulated Chemokine 1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CXCL17/VCC-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse CXCL17 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction