Learn More
Human CXCL9/MIG Alexa Fluor 647-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC392R-100UG
This item is not returnable.
View return policy
Description
CXCL9/MIG Monoclonal specifically detects CXCL9/MIG in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| CXCL9/MIG | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q07325 | |
| CXCL9 | |
| E. coli-derived recombinant human CXCL9/MIG Thr23-Thr125 Accession # Q07325 | |
| RUO | |
| 4283 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 49106 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, Gamma-interferon-induced monokine, Humig, MIG, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human CXCL9/MIG in ELISAs and Western blots. In ELISAs, does not cross-react with recombinant mouse (rm) CXCL9, recombinant human CXCL10. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Product Title
For Research Use Only (RUO)