missing translation for 'onlineSavingsMsg'
Learn More

Human CXCR7/RDC-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  R&D Systems FAB42273G-100UG

Product Code. 18731549

  • ONLINE EXCLUSIVE PRICE
    € 461.00
    € 435.75 / 100µg
    Save € 25.25
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CXCR7/RDC-1
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P25106
CXCR7
Human CXCR7 encoding plasmid Accession # P25106
RUO
57007
Human
IgG1
Flow Cytometry
11G8
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog
Mouse
100 μg
Primary
Detects human CXCR7/RDC-1 in flow cytometry of five distinct human CXCR7 transfectants, but not their respective parental lines.
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only (RUO)