missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CYB561 (aa 221-247) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53228 (PA5-53228. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Secretory vesicle-specific electron transport protein.
Specifications
Specifications
| Accession Number | P49447 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 1534 |
| Name | Human CYB561 (aa 221-247) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | CD158F; CD158F1; Cyb561; CYB561A1; cytochrome b561; Cytochrome b-561; cytochrome b561 family, member A1; ferric-chelate reductase 2; FRRS2; KIR2DL5; Mcyt |
| Common Name | CYB561 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction