missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CYB5B (aa 19-112) Control Fragment Recombinant Protein

Product Code. 30213328
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30213328 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30213328 Supplier Invitrogen™ Supplier No. RP89712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52482 (PA5-52482. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CYB5B gene ontology annotations related to this gene include enzyme activator activity; heme binding; metal ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43169
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80777
Name Human CYB5B (aa 19-112) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1810044O22Rik; AU015618; Cyb5b; Cyb5m; CYB5-M; CYPB5M; cytochrome b5 outer mitochondrial membrane isoform; Cytochrome b5 type B; cytochrome b5 type B (outer mitochondrial membrane); cytochrome b5, outer mitochondrial membrane isoform; OMB5; outer mitochondrial membrane cytochrome b5; type 2 cyt-b5
Common Name CYB5B
Gene Symbol CYB5B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EVETSVTYYRLEEVAKRNSLKELWLVIHGRVYDVTRFLNEHPGGEEVLLEQAGVDASESFEDVGHSSDAREMLKQYYIGDIHPSDLKPESGSKD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.