missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Cynomolgus Monkey FCAR/CD89 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3939R-100UG
This item is not returnable.
View return policy
Description
FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Human, Cynomolgus Monkey samples. It is validated for Flow Cytometry.
Specifications
| FCAR/CD89 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P24071 | |
| FCAR | |
| NS0 mouse myeloma cell line transfected with human FCAR/CD89 Gln22-Lys287 Accession # P24071 | |
| RUO | |
| 2204 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 488032 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD89, CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FCAR/CD89 in direct ELISAs. In direct ELISAs, no cross-reactivity with Fc gamma RIA, RIIA, or RIIIB is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction