missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin A Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF1407
This item is not returnable.
View return policy
Description
Cystatin A Polyclonal specifically detects Cystatin A in Human samples. It is validated for Western Blot.
Specifications
| Cystatin A | |
| Polyclonal | |
| Biotin | |
| P01040 | |
| CSTA | |
| E. coli-derived recombinant human Cystatin A Ile2-Phe98 Accession # P01040 | |
| 50 μg | |
| Primary | |
| Detects human Cystatin A in Western blots. In Western blots, less than 2% cross-reactivity with recombinant human (rh) Cystatin B, rhCystatin C, rhCystatin D, rhCystatin E/M, rhCystatin F, rhCystatin S, rhCystatin SA and rhCystatin SN is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| AREI, CSTA, cystatin A (stefin A), cystatin AS, cystatin-A, Stefin A, STF1, STF1Cystatin-AS, STFA, STFAStefin-A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1475 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction