missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytokeratin 18 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC7619G-100UG
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human samples. It is validated for Immunocytochemistry
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, Keratin 18, keratin, type I cytoskeletal 18, keratin-18, KRT18 | |
| E. coli-derived recombinant human Cytokeratin 18, Ala239-Asp397, Accession # P05783 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 810811 | |
| Immunocytochemistry 1:10 dilution | |
| P05783 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3875 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction