missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytokeratin 7 Antibody, R&D Systems™
Mouse Monoclonal Antibody
€ 228.00 - € 561.00
Specifications
| Antigen | Cytokeratin 7 |
|---|---|
| Clone | 1090519 |
| Dilution | Western Blot 0.5 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279864
|
R&D Systems
MAB11626-SP |
25 μg |
€ 228.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279872
|
R&D Systems
MAB11626-100 |
100 μg |
€ 561.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Immunocytochemistry,Western Blot,ImmunohistochemistrySpecifications
| Cytokeratin 7 | |
| Western Blot 0.5 μg/mL, Immunohistochemistry 3-25 μg/mL, Immunocytochemistry 3-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| 3855 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
| 1090519 | |
| Immunocytochemistry, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7 | |
| Synthetic Peptide | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title