missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cytosolic Sulfotransferase 2A1/SULT2A1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5828
This item is not returnable.
View return policy
Description
Cytosolic Sulfotransferase 2A1/SULT2A1 Polyclonal specifically detects Cytosolic Sulfotransferase 2A1/SULT2A1 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| Cytosolic Sulfotransferase 2A1/SULT2A1 | |
| Polyclonal | |
| Unconjugated | |
| Q06520 | |
| SULT2A1 | |
| E. coli-derived recombinant human Cytosolic Sulfotransferase 2A1/SULT2A1 Ser2-Glu285 Accession # Q06520 | |
| 100 μg | |
| Primary | |
| Detects human Cytosolic Sulfotransferase 2A1/SULT2A1 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) SULT1A1, rhSULT1A3, rhSULT1B1, rhSULT1E1, and rhSULT4A1 is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Neutralization 35.9 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| alcohol/hydroxysteroid sulfotransferase, Dehydroepiandrosterone sulfotransferase, DHEAS, DHEA-ST, DHEA-STbile salt sulfotransferase, EC 2.8.2.14, HST, hSTa, Hydroxysteroid Sulfotransferase, ST2, ST2A1, ST2A3, STDbile-salt sulfotranasferase 2A1, Sulfotransferase 2A1, sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone(DHEA)-preferring, member 1, SULT2A1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6822 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction