Learn More
Human Cytosolic Sulfotransferase 2B1/SULT2B1 Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  R&D Systems AF6174
Product Code. 18028471
  • € 501.00 / 100µg
Description
Cytosolic Sulfotransferase 2B1/SULT2B1 Polyclonal specifically detects Cytosolic Sulfotransferase 2B1/SULT2B1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytosolic Sulfotransferase 2B1/SULT2B1
Polyclonal
Unconjugated
O00204
SULT2B1
E. coli-derived recombinant human Cytosolic Sulfotransferase 2B1/SULT2B1 isoform 1 Asp2-Ser365 Accession # O00204
100 μg
Primary
Detects human SULT2B1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) SULT1B1, rhSULT1A1, rhSULT2A1, rhSULT4A1, and rhSULT1E1 is observed.
Human
IgG
Western Blot, Immunohistochemistry, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Alcohol sulfotransferase, EC 2.8.2, EC 2.8.2.2, HSST2, HSST2sulfotransferase family cytosolic 2B member 1, Hydroxysteroid sulfotransferase 2, ST2B1, Sulfotransferase 2B1, sulfotransferase family, cytosolic, 2B, member 1, SULT2B1
Sheep
Affinity Purified
RUO
6820
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.