missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAB2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8064
This item is not returnable.
View return policy
Description
DAB2 Monoclonal specifically detects DAB2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| DAB2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P98082 | |
| DAB2 | |
| E. coli-derived recombinant human DAB2 Lys630-Ala770 Accession # P98082 | |
| 100 μg | |
| Primary | |
| Detects human DAB2 in ELISA and Western Blot. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 883216 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Differentially-expressed protein 2, disabled (Drosophila) homolog 2 (mitogen-responsive phosphoprotein), disabled homolog 2, mitogen-responsive phosphoprotein (Drosophila), Disabled-2, DOC2, DOC-2DOC2disabled homolog 2, FLJ26626, mitogen-responsive phosphoprotein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1601 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction