missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DARC Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB4139R-100UG
This item is not returnable.
View return policy
Description
DARC Monoclonal specifically detects DARC in Human samples. It is validated for Flow Cytometry.
Specifications
| DARC | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q16570 | |
| DARC | |
| HEK293 human embryonic kidney cell line transfected with human DARC Met1-Ser336 Accession # Q16570 | |
| RUO | |
| 2532 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 358307 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ACKR1, CCBP1, CD234, CD234 antigen, Dfy, Duffy Antigen, Duffy antigen/chemokine receptor, Duffy blood group antigen, Duffy blood group, chemokine receptor, Fy Antigen, Fy glycoprotein, FYDuffy blood group, Glycoprotein D, GPD, GPDWBCQ1, GpFy, Plasmodium vivax receptor | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human DARC. Stains human DARC transfected cells but not irrelevant transfectants. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction