missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DC-LAMP Lumenal Domain Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4087
This item is not returnable.
View return policy
Description
DC-LAMP Polyclonal specifically detects DC-LAMP in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| DC-LAMP | |
| Polyclonal | |
| Unconjugated | |
| EAW78337 | |
| LAMP3 | |
| Mouse myeloma cell line NS0-derived recombinant human DC-LAMP Lumenal Domain Asp21-Thr381 Accession # EAW78337 | |
| 100 μg | |
| Primary | |
| Detects human DC-LAMP Lumenal Domain in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human LAMP is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CD208, CD208 antigen, DCLAMP, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP3, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403, TSC403CD208 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27074 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction